Orders LinkContactUsLinkComponent
Wire basket, rectangular
Wire basket, rectangular
430×310×220 mm
Catalog # SEMA2659
Supplier:   SEMADENI
Wire basket, rectangular
Catalog # SEMA2659
Supplier:  SEMADENI
Wire basket, rectangular
430×310×220 mm
Price
39,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of PP, anthracite.
  • Good chemical and temperature resistance
  • Space-saving stackable
  • With stable, foldable plastic support bracket with locking in vertical position
Specifications
  • W×D×H:
    430×310×220 mm