DNA methylation kit, express
Supplier: Enzo Life Sciences
Certificates
About this item
Express DNA Methylation Kit can be used for rapid and reliable bisulfite treatment and conversion of DNA for methylation analysis.
- High yield and ready to use
- Easy to follow instructions
Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Delivery: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, Methylation elution buffer , IC column, IC collection tube and L-Desulphonation reagent.
Frequently Bought Together