Purge and trap spargers
Supplier: Restek
Certificates
About this item
These purge and trap spargers are designed for use with Tekmar purge-and-trap instruments only and manufactured to tight tolerances to ensure a leak-tight seal.
- Available with uniform frits to ensure maximum purging efficiency
- Non-fritted spargers can be used for wastewater samples
- With 12,7 mm mount
Frequently Bought Together