Orders LinkContactUsLinkComponent
Modules for inoLab® Multi IDS
Supplier:   WTW
Modules for inoLab® Multi IDS
Supplier:  WTW
Certificates
Modules for inoLab® Multi IDS
Modules for inoLab® Multi IDS
2 Options Available Below
Product Details & Documents
Integratable adapters for connecting Multi IDS meters with conventional pH/ORP/ISE electrodes.
Product Family Options
Product Information
  • Availability
    Price
  • 665-0371
    ADA 94pH/IDS DIN, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with DIN- and 4 mm banana plug, incl. mounting accessories
    ADA 94pH/IDS DIN, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with DIN- and 4 mm banana plug, incl. mounting accessories
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 405,00 €
    405,00 €
    1 * 1 ST
       405,00 €
  • 665-0372
    ADA 94pH/IDS BNC, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with BNC- and 4 mm banana plug, incl. mounting accessories
    ADA 94pH/IDS BNC, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with BNC- and 4 mm banana plug, incl. mounting accessories
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 405,00 €
    405,00 €
    1 * 1 ST
       405,00 €