Order Entry
Belgium
ContactUsLinkComponent
Ductless filtering fume hoods, Captair® Smart 633
Ductless filtering fume hoods, Captair® Smart 633
  ERLAAA656A00011A21
New Product
Ductless filtering fume hoods, Captair® Smart 633
  ERLAAA656A00011A21
New Product

 

  • W×D×H ext.:
    1600×750×1340/1515 mm
  • Description:
    Captair® 633 Smart, EU-plug
  • Model:
    Captair® Smart 633
  • Air face velocity:
    0,4 −0,6
  • Energy consumption:
    160 W
  • Electrical:
    220 V/50-60 Hz
  • Airflow:
    660 m3 - 135 cfm
  • Lighting:
    LED>650 lux
  • Specifications:
    Sash opening: total openings or new reverso sash
  • Pk:
    1

 

 

The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://uk.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=20532418

  • A simple light pattern and sound pulses indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough
  • Felxibility: Modular filtration column adapting to application changes
  • Real time sensor to detect main filter satuation with solvents, acids and formaldehyde
  • No chemical release into the atmosphere
  • No ductwork cost
  • Low power consumption
  • New electronic anemometer monitors face velocity
  • The 6° slanted sash offers optimal visibility and ergonomic work space
  • Large openings for easy access to the work

The structure is made from corrosion resistant electro-galvanised steel coated with anti-acid polymer, sides and front panels are in chemical resistant acrylic.

The configurable filtering system above the enclosure can be adapted to filter gases, solvents, powders and particulates keeping the user and the laboratory protected by safely and efficiently recirculating air within the room and releasing purified air back into the room free of any toxic hazardous vapours or odours.

The monitoring gives the user the control to monitor the hood, change the settings and delivers safety alerts immediately to the mobile, tablet or PC device.

Types of carbon filters:

AS: For organics vapours
BE: Polyvalent for acid and organic vapours
F: For formaldehyde vapours
K: For ammonia vapours

Filtration fault detection: Detection sensors for solvents, acids or formaldehyde.
Monitoring for ambient handling conditions: Temperature and Hygrometry sensors.

 : Meets AFNOR NFX 15 211/ANSI Z9.5-2012 filtration efficiency standard (classes 1 and 2).
 : There is a very wide range of Erlab Fume Hoods and Enclosures. To ensure you receive the correct product for your specific application and setting please complete the contact form as per the url above. Your local Avantor sales representative will then send you an eValiQuest form to get details of your application and requirements. Once we have received this back from you, we will send you a quotation based on your precise requirements ensuring all handling and safety requirements are met.