Orders LinkContactUsLinkComponent
Stepladder, non-mobile
Non-mobile step ladder, 2, 805×500×600 mm
  PALB2-LAD-NM
 :   Palbam
Stepladder, non-mobile
  PALB2-LAD-NM
 :  Palbam
Stepladder, non-mobile
Stepladder, non-mobile
Non-mobile step ladder, 2, 805×500×600 mm
1 * 1 ST
These step stools are designed for fixed locations where they are not required to be moved often.
  • Made up of 304 stainless steel with electropolished finish
  • No. of steps:
    2
  • Description:
    Non-mobile step ladder
  • W×D×H:
    805×500×600 mm