Orders LinkContactUsLinkComponent
Caps for screw thread vials
Supplier:   Restek
Caps for screw thread vials
Supplier:  Restek
Certificates
Caps for screw thread vials
Caps for screw thread vials
2 Options Available Below
Product Details & Documents
Black PP, open hole caps fit all 2,0 ml, 12×32 mm, screw-thread 8 mm/425 vial-based autosamplers.
Product Family Options
Product Information
  • Availability
    Price
  • REST21176
    Open-hole caps, PP, black
    Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 14,50 €
    14,50 €
    1 * 100 ST
       14,50 €
  • REST21177
    Open-hole caps, PP, black
    Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 94,80 €
    94,80 €
    1 * 1.000 ST
       94,80 €