GC ferrules, Graphite Vespel®
: Restek
These ferrules can be used with Vacuum Vu-Union® connectors.
- Made of 60% Vespel® and 40% graphite
- Maximum operating temperature up to 400 °C
Frequently Bought Together
These ferrules can be used with Vacuum Vu-Union® connectors.