Orders LinkContactUsLinkComponent
Screw thread vials, 8-425
Screw thread vials, 8-425
Supplier:   Restek
Screw thread vials, 8-425
Supplier:  Restek
Certificates
Screw thread vials, 8-425
Screw thread vials, 8-425
3 Options Available Below
Product Details & Documents
Screw thread vials fits all 2,0 ml, 12×32 mm vial based autosamplers and provide most secure environment for your sample because evaporation is minimised.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Availability
    Price
  • REST24619
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Clear
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 40,20 €
    40,20 €
    1 * 100 ST
       40,20 €
  • REST24620
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Clear
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 323,00 €
    323,00 €
    1 * 1.000 ST
       323,00 €
  • REST24622
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Amber
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 369,00 €
    369,00 €
    1 * 1.000 ST
       369,00 €
Caps
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 14,50 €
    14,50 €
    1 * 100 ST
       14,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 94,80 €
    94,80 €
    1 * 1.000 ST
       94,80 €
Septa
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/white silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 38,20 €
    38,20 €
    1 * 100 ST
       38,20 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/silicone
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 255,00 €
    255,00 €
    1 * 1.000 ST
       255,00 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/white silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 264,00 €
    264,00 €
    1 * 1.000 ST
       264,00 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/silicone
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 36,10 €
    36,10 €
    1 * 100 ST
       36,10 €
Caps and septa (preassembled)
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    Red PTFE/silicone, 1,65 mm
    8 mm
    Caps and septa, preassembled
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 47,40 €
    47,40 €
    1 * 100 ST
       47,40 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    PTFE/silicone
    8 mm
    Caps and septa, preassembled, top hat
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,80 €
    61,80 €
    1 * 100 ST
       61,80 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    Red PTFE/silicone, 1,65 mm
    8 mm
    Caps and septa, preassembled
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 401,00 €
    401,00 €
    1 * 1.000 ST
       401,00 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    PTFE/silicone
    8 mm
    Caps and septa, preassembled, top hat
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 523,00 €
    523,00 €
    1 * 1.000 ST
       523,00 €