ContactUsLinkComponent
Filter plates, 96-well, with long tip filter, for nucleic acid binding (NAB), AcroPrep™ Advance
Filter plates, 96-well, with long tip filter, for nucleic acid binding (NAB), AcroPrep™ Advance
Catalog # 738-0209
undefined
Filter plates, 96-well, with long tip filter, for nucleic acid binding (NAB), AcroPrep™ Advance
Catalog # 738-0209
Specifications
  • Membrane:
    Glass fibre
  • No. of wells:
    96
  • Colour:
    Natural
  • Description:
    AcroPrep™ Advance, long tip filter
  • Pk:
    5
  • Well volume:
    1 ml
Specifications
About this item
The Pall AcroPrep™ Advance 96-well long tip filter plate for nucleic acid binding (NAB) incorporates a silica-based quartz glass fibre media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples. This media offers researchers the flexibility to purify plasmid DNA from bacteria, and genomic DNA or total RNA from cell culture samples using a single plate.
  • Single plate for multiple applications reduces chance of cross contamination - critical for reproducible, high quality results
  • Single plate aids smooth sample processing flow
  • Long tip design minimises hanging drop formation