Handles for microsurgical blades, Feather®
Supplier: PFM Medical GmbH
Certificates
About this item
These curved and bayonet-shaped microsurgical handles feature a unique design. They can be used with the different microsurgical blades.
- Made from titanium alloy
- Ideal weight ensures a secure grip and safe handling of the instrument
Frequently Bought Together