Scalpels, disposable, Feather®
Supplier: PFM Medical GmbH
Certificates
About this item
These disposable standard scalpels with high-quality stainless steel blade are suitable for all surgical applications.
- Outstanding quality and performance thanks to optimised micrometer-accurate precision grinding
- Double-honing process ensures ultra-sharp cutting edges of great durability
Packaging: 20 pieces per pack.
Frequently Bought Together