Order Entry
Belgium
Orders LinkContactUsLinkComponent
Utility tray

Utility tray

Utility tray, holds 102 l of spilled liquid, Black, 1020×1020×130 mm
Catalog # NEWPPAK922
Supplier:   NEW PIG
Utility tray
Catalog # NEWPPAK922
Supplier:  NEW PIG
Utility tray
Utility tray

Utility tray, holds 102 l of spilled liquid, Black, 1020×1020×130 mm

Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This durable, one-piece utility trays contain liquid messes from oily parts and leaky containers.

  • Thick-wall, chemical-resistant polyethylene construction won't bend or warp
  • Ribs on bottom of tray add strength and elevate parts to keep them away from trapped liquids
  • Ideal for temporary storage of small containers that may leak or drip liquids onto the floor
Specifications
  • Colour:
    Black
  • Description:
    Utility tray, holds 102 l of spilled liquid
  • W×D×H:
    1020×1020×130 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.