ContactUsLinkComponent
FID/NPD adapter fitting
FID/NPD adapter fitting
Catalog # REST20884
Supplier:  Restek
undefined
FID/NPD adapter fitting
Catalog # REST20884
Supplier:  Restek
Specifications
  • Description:
    FID/NPD adaptor fitting
  • Pk:
    1
Specifications
About this item
This easy to use adapter fitting is sturdy and made of compact stainless steel.
  • ¹/₁₆" nut uses standard graphite or vespel/graphite ferrules
  • Wrench pad does not turn when installing a capillary column
Ordering information: Supplied with 1/4 and 1/16" stainless steel nuts, and 1/4" vespel and 0,4 mm ID graphite ferrules.