ContactUsLinkComponent
Injector/detector plug nuts
Injector/detector plug nuts
Catalog # REST21883
Supplier:  Restek
undefined
Injector/detector plug nuts
Catalog # REST21883
Supplier:  Restek
Specifications
  • Description:
    Injector/detector plug nuts
  • Pk:
    2
Specifications
About this item
These are used to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base.
  • Used to cap off an injector to isolate leaks and a detector for thermal cleaning
  • To check a detector or make-up gas flow rate