FID collector assembly kit
Fournisseur: Restek
Certificats
À propos de cet article
These FID collector assembly kits are made of high-quality stainless steel. Ideal for Agilent 5890/6850/6890/7890 GCs.
Frequently Bought Together
These FID collector assembly kits are made of high-quality stainless steel. Ideal for Agilent 5890/6850/6890/7890 GCs.