Orders LinkContactUsLinkComponent
Wafer tweezers
Wafer tweezers
Supplier:   OUTILS RUBIS
Wafer tweezers
Supplier:  OUTILS RUBIS
Certificates
Wafer tweezers
2 Options Available Below
Product Details & Documents
Stainless steel. These narrow tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddel for glare prevention
  • Each Wafer size has its corresponding tweezers size (e.g. for an 8" Wafer take tweezers with a No. 8)
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • OUTI37S-1
    Without teeth, narrow (width 3 mm), flat lower paddle
    Without teeth, narrow (width 3 mm), flat lower paddle
    37S-1
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,30 €
    50,30 €
    1 * 1 ST
       50,30 €
  • OUTI37S-2
    Without teeth, narrow (width 4,5 mm), flat lower paddle
    Without teeth, narrow (width 4,5 mm), flat lower paddle
    37S-2
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,50 €
    57,50 €
    1 * 1 ST
       57,50 €