Orders LinkContactUsLinkComponent
Multipacks, Tacta®
   Sustainable Options Available
Supplier:   Sartorius
Multipacks, Tacta®
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Multipacks, Tacta®
8 Options Available Below
Product Details & Documents
The Tacta® Multipacks offer an opportunity to test and get started with Tacta® mechanical pipettes. Tacta® mechanical pipette offers excellent performance and safety in manual pipetting. It is designed to maintain high accuracy and precision in repetitive and long-lasting pipetting series.
  • Reliable results, even over lengthy pipetting periods
  • Controlled and smooth tip ejection
  • The Optilock® system provides flexibility for volume adjustment and locking
There are different multipack options to choose from ranging from a set of 3 to a set of 5 pipettes.
 : All sets come with a linear stand for the safe storage of your pipettes as well as suitable racked or bulk-packed Optifit tips.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-5626
  • 613-5627
  • 613-5628
  • 613-5629
  • 613-5630
  • 613-5631
  • 613-5632
  • 613-5633
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 613-5626
    3-pack 10, incl. three single channel pipettes (0,5 - 10 / 10 - 100 / 100 - 1000 µl), Optifit tip tray boxes (0,2 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    3-pack 10, incl. three single channel pipettes (0,5 - 10 / 10 - 100 / 100 - 1000 µl), Optifit tip tray boxes (0,2 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 964,00 €
    964,00 €
    1 * 1 Un.
       964,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5626
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5627
    3-pack 20, incl. three single channel pipettes (2 - 20 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    3-pack 20, incl. three single channel pipettes (2 - 20 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 964,00 €
    964,00 €
    1 * 1 Un.
       964,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5627
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5628
    4-pack 20, incl. four single channel pipettes (0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    4-pack 20, incl. four single channel pipettes (0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 1 260,00 €
    1 260,00 €
    1 * 1 Un.
       1 260,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5628
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5629
    4-pack 100, incl. four single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    4-pack 100, incl. four single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 1 260,00 €
    1 260,00 €
    1 * 1 Un.
       1 260,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5629
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5630
    5-pack 10, incl. five single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 / 500 - 5000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 / 100 - 5000 μl; 96 tips each) and one linear stand
    5-pack 10, incl. five single channel pipettes (0,5 - 10 / 10 - 100 / 20 - 200 / 100 - 1000 / 500 - 5000 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 10 - 1000 / 100 - 5000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 1 570,00 €
    1 570,00 €
    1 * 1 Un.
       1 570,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5630
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5631
    5-pack 20, incl. five single channel pipettes (2 - 20 / 10 - 100 / 20 - 200 / 100 - 1000 / 500 - 5000 µl), Optifit tip tray boxes (0,5 - 200 / 10 - 1000 / 100 - 5000 μl; 96 tips each) and one linear stand
    5-pack 20, incl. five single channel pipettes (2 - 20 / 10 - 100 / 20 - 200 / 100 - 1000 / 500 - 5000 µl), Optifit tip tray boxes (0,5 - 200 / 10 - 1000 / 100 - 5000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 1 570,00 €
    1 570,00 €
    1 * 1 Un.
       1 570,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5631
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5632
    3+1-pack, incl. three single channel pipettes (0,5 - 10 / 20 - 200 / 100 - 1000 µl) and one 8-channel pipette (30 - 300 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 5 - 350 / 10 - 1000 μl; 96 tips each) and one linear stand
    3+1-pack, incl. three single channel pipettes (0,5 - 10 / 20 - 200 / 100 - 1000 µl) and one 8-channel pipette (30 - 300 µl), Optifit tip tray boxes (0,1 - 10 / 0,5 - 200 / 5 - 350 / 10 - 1000 μl; 96 tips each) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 1 740,00 €
    1 740,00 €
    1 * 1 Un.
       1 740,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5632
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • 613-5633
    3-pack MAX, incl. three single channel pipettes (100 - 1000 / 500 - 5000 / 1000 - 10000 µl), Optifit tips (10 - 1000 μl (96/tray) / 0,1 - 5 ml, bulk (100) / 1 - 10 ml, bulk (50)) and one linear stand
    3-pack MAX, incl. three single channel pipettes (100 - 1000 / 500 - 5000 / 1000 - 10000 µl), Optifit tips (10 - 1000 μl (96/tray) / 0,1 - 5 ml, bulk (100) / 1 - 10 ml, bulk (50)) and one linear stand
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un. - 964,00 €
    964,00 €
    1 * 1 Un.
       964,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5633
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact