ContactUsLinkComponent
VWR® PESTINORM®, Dimethylsulphoxide (DMSO), Secondary Reference Standard for GC
VWR® PESTINORM®, Dimethylsulphoxide (DMSO), Secondary Reference Standard for GC
  85663.180
 :  
undefined
VWR® PESTINORM®, Dimethylsulphoxide (DMSO), Secondary Reference Standard for GC
  85663.180
 :  
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Dimethylsulphoxide (DMSO) standard for GC
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID