ContactUsLinkComponent
Open end wrench set
Open end wrench set
Catalog # REST20387
Supplier:  Restek
undefined
Open end wrench set
Catalog # REST20387
Supplier:  Restek
Specifications
  • Description:
    Open end wrench set
  • Pk:
    1
Specifications
About this item
High-quality 1/4×5/16", 3/8"×7/16", 7/16"×1/2", and 1/2"×9/16" wrenches for tightening a wide variety of chromatography fittings.