Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cell lysis buffer, RIPA

Cell lysis buffer, RIPA

RIPA cell lysis buffer 3, 7,4, 100 mL
Catalog # ENZOADI801496
Supplier:   Enzo Life Sciences
Cell lysis buffer, RIPA
Catalog # ENZOADI801496
Supplier:  Enzo Life Sciences
CAS Number:  
Cell lysis buffer, RIPA

RIPA cell lysis buffer 3, 7,4, 100 mL

CAS Number:  
Price
259,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

The RIPA liquid cell lysis buffer composes of 50 mM TRIS-hydrogen chloride, pH 7,4, containing 150 mM sodium chloride, 1 mM EDTA, 1 mM EGTA, 1,2% Triton X-100, 0,5% sodium deoxycholate, and 0,1% SDS.

Storage temperature: 4 °C

Caution: RUO - Research use only.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    RIPA cell lysis buffer 3
  • pH value:
    7,4
  • Cat. No.:
    ENZOADI801496
  • Formulation:
    Liquid. 50 mM Tris, containing 150 mM NaCl, 1 mM EDTA, 1 mM EGTA, 1,2% Tergitol 15-S-9, 0,5% sodium deoxycholate, and 0,1% SDS
  • Storage temperature:
    4 °C (Long term storage)
  • Shipping conditions:
    20

Specifications