GC columns, ShinCarbon ST (packed and micropacked)
Supplier: Restek
Certificates
About this item
These ShinCarbon ST columns are made of SilcoSmooth stainless steel material. Analyses oxygen, nitrogen, methane, carbon monoxide and carbon dioxide with one column at room temperature and separate permanent gases, including carbon monoxide and carbon dioxide, without cryogenic cooling.
- Rapid separations of permanent gas/light hydrocarbon mixtures
- Compatibile with most GC detectors - minimal bleed, minimal baseline rise
- Preconditioned, less than 30 minutes to stabilise
- Maximum temperature of 280/300 °C
ShinCarbon ST material, a high surface area carbon molecular sieve (~1500 m²/g), is the ideal medium for separating gases and highly volatile compounds by gas solid chromatography (GSC). Low-bleed operation and rapid stabilisation. Its 300 °C upper programmed temperature limit minimises bleed and baseline rise during temperature programming, making the material compatible with most detection systems used for gas analysis, including TCD or HID.
Ordering information: Columns not include column nuts and ferrules. Optional installation kits can be ordered separately.
Specifications
- Catalog No:
- REST80486-810
- REST80486-820
- REST19043
- REST19809
- REST19808
- REST80486-800
- REST80486-840
- REST19810
- Temperature limit:
- 300 °C
- up to 300 °C
- up to 300 °C
- up to 300 °C