Precision sampling valves for vials, Mininert®
Supplier: Restek
Jump to:
About this item
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
8 Options Available Below
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
Product Family Options
Product Information
- AvailabilityPrice
- REST24900Mininert® precision sampling valves, for screw-thread vials, 13 mm/425 threadMininert® precision sampling valves, for screw-thread vials, 13 mm/425 threadLog in for availability1 * 12 ST236,00 €
- REST24901Mininert® precision sampling valves, for screw-thread vials, 15 mm/425 threadMininert® precision sampling valves, for screw-thread vials, 15 mm/425 threadLog in for availability1 * 12 ST232,00 €
- REST24902Mininert® precision sampling valves, for screw-thread vials, 18 mm/400 threadMininert® precision sampling valves, for screw-thread vials, 18 mm/400 threadLog in for availability1 * 12 ST254,00 €
- REST24903Mininert® precision sampling valves, for screw-thread vials, 20 mm/400 threadMininert® precision sampling valves, for screw-thread vials, 20 mm/400 threadLog in for availability1 * 12 ST267,00 €
- REST24904Mininert® precision sampling valves, for screw-thread vials, 24 mm/400 threadMininert® precision sampling valves, for screw-thread vials, 24 mm/400 threadLog in for availability1 * 12 ST278,00 €
- REST24905Mininert® precision sampling valves, for crimp-top vials
This product has been blocked by your organization. Please contact your purchasing department for more information
Accessories