Extraction cell bodies for solvent extraction, ASE® systems
Fournisseur: Restek
Certificats
À propos de cet article
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 method 3545 and pressurised fluid extraction (PFE).
- Cell bodies are serialised for easy sample identification
- Smooth inner surfaces for easier cleaning
Informations de commande: Caps are sold separately.
Frequently Bought Together