Capillary columns, Stx-CLPesticides (fused silica with siltek deactivation)
Supplier: Restek
Certificates
About this item
These application-specific columns with proprietary crossbond phases are suitable for organochlorine pesticides and herbicides.
- Baseline separations in less than 10 minutes
- Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
- Temperature range stable to 330 °C
Specifications
- Catalog No:
- REST11543
- REST11444-600
- REST11544
- REST11440
- REST11441
- REST11544-600
- REST11540
- REST11541
- REST11443
- REST11444
- Temperature limit:
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C
- −60...+310/330 °C