Anti-SUMO1 Rabbit Polyclonal Antibody
Catalog # ABGEAP1222E
Supplier: Abgent
Specifications
- Antigen name:Small ubiquitin-related modifier 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Epitope:C-Terminal D86
- Antigen synonyms:Sentrin|UBL1|Ubiquitin-like protein UBL1|SMT3H3|GAP modifying protein 1|SMT3C|SUMO-1|SMT3 homolog 3|GMP1|Small ubiquitin-related modifier 1|Ubiquitin-like protein SMT3C|Ubiquitin-homology domain protein PIC1
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type:
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: C-Terminal D86
Host: Rabbit
Isotype:
Reactivity: