Order Entry
Belgium
ContactUsLinkComponent
Anti-TFRC Mouse Monoclonal Antibody (CF647) [clone: TFRC/1149]
Anti-TFRC Mouse Monoclonal Antibody (CF647) [clone: TFRC/1149]
Supplier:  Biotium
Certificates
Anti-TFRC Mouse Monoclonal Antibody (CF647) [clone: TFRC/1149]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC471149-100
  • BNC471149-500
  • Antigen symbol:
  • TFRC
  • TFRC
  • Antigen name:
  • CD71
  • CD71
  • Antigen synonyms:
  • TRFR|TFRC transferrin receptor (p90 CD71)|TFR1|Mtvr-1|p90
  • TRFR|TFRC transferrin receptor (p90 CD71)|TFR1|Mtvr-1|p90
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone TFRC/1149
  • Clone TFRC/1149
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 529618
  • 529618
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together