About this item
This antibody recognizes a ~90-95 kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190 kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: TFRC/1149
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC941149-100
- BNC941149-500
- Antigen symbol:
- TFRC
- TFRC
- Antigen name:
- CD71
- CD71
- Antigen synonyms:
- TRFR|TFRC transferrin receptor (p90 CD71)|TFR1|Mtvr-1|p90
- TRFR|TFRC transferrin receptor (p90 CD71)|TFR1|Mtvr-1|p90
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone TFRC/1149
- Clone TFRC/1149
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 529618
- 529618
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide