Order Entry
Belgium
ContactUsLinkComponent
Anti-ZAP70 Mouse Monoclonal Antibody (CF647) [clone: ZAP70/528 + 2F3.2]
Anti-ZAP70 Mouse Monoclonal Antibody (CF647) [clone: ZAP70/528 + 2F3.2]
Supplier:  Biotium
Certificates
Anti-ZAP70 Mouse Monoclonal Antibody (CF647) [clone: ZAP70/528 + 2F3.2]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC471137-500
  • BNC471137-100
  • Antigen symbol:
  • ZAP70
  • ZAP70
  • Antigen synonyms:
  • Selective T cell defect|SRK|Tyrosine-protein kinase ZAP-70|Zeta chain associated protein kinase 70kDa|TZK|Syk-related tyrosine kinase|STD
  • Selective T cell defect|SRK|Tyrosine-protein kinase ZAP-70|Zeta chain associated protein kinase 70kDa|TZK|Syk-related tyrosine kinase|STD
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone ZAP70/528 + 2F3.2
  • Clone ZAP70/528 + 2F3.2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 234569
  • 234569
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together