Order Entry
Belgium
ContactUsLinkComponent
Anti-ACP5 Mouse Monoclonal Antibody (CF488A) [clone: ACP5/1070]
Anti-ACP5 Mouse Monoclonal Antibody (CF488A) [clone: ACP5/1070]
Supplier:  Biotium
Certificates
Anti-ACP5 Mouse Monoclonal Antibody (CF488A) [clone: ACP5/1070]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881070-100
  • BNC881070-500
  • Antigen symbol:
  • ACP5
  • ACP5
  • Antigen name:
  • TRAcP
  • TRAcP
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone ACP5/1070
  • Clone ACP5/1070
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 1211
  • 1211
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together