Anti-ITGA2B Mouse Monoclonal Antibody (CF640R) [clone: ITGA2B/1036]
Supplier: Biotium
Certificates
About this item
Reacts with a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: ITGA2B/1036
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC401036-500
- BNC401036-100
- Antigen symbol:
- ITGA2B
- ITGA2B
- Antigen name:
- CD41a
- CD41a
- Antigen synonyms:
- Integrin alpha IIb|platelet fibrinogen receptor|alpha subunit|HPA3|BDPLT2|platelet glycoprotein IIb of IIb/IIIa complex|ITGAB|BDPLT16|GT|ITGA2B|GPalpha IIb
- Integrin alpha IIb|platelet fibrinogen receptor|alpha subunit|HPA3|BDPLT2|platelet glycoprotein IIb of IIb/IIIa complex|ITGAB|BDPLT16|GT|ITGA2B|GPalpha IIb
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone ITGA2B/1036
- Clone ITGA2B/1036
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 411312
- 411312
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together