Order Entry
Belgium
ContactUsLinkComponent
Anti-MITF Mouse Monoclonal Antibody (CF640R) [clone: MITF/915]
Anti-MITF Mouse Monoclonal Antibody (CF640R) [clone: MITF/915]
Supplier:  Biotium
Certificates
Anti-MITF Mouse Monoclonal Antibody (CF640R) [clone: MITF/915]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400915-100
  • BNC400915-500
  • Antigen symbol:
  • MITF
  • MITF
  • Antigen name:
  • MiTF
  • MiTF
  • Antigen synonyms:
  • CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
  • CMM8|BHLHE32|MITF|Class E basic helix-loop-helix protein 32 (bHLHe32)|WS2|WS2A|Microphthalmia-associated transcription factor|Mi
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone MITF/915
  • Clone MITF/915
  • Reactivity:
  • Human, Dog
  • Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 166017 , 618266
  • 166017 , 618266
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together