Order Entry
Belgium
ContactUsLinkComponent
Anti-HSPD1 Mouse Monoclonal Antibody (CF488A) [clone: HSPD1/780]
Anti-HSPD1 Mouse Monoclonal Antibody (CF488A) [clone: HSPD1/780]
Supplier:  Biotium
Certificates
Anti-HSPD1 Mouse Monoclonal Antibody (CF488A) [clone: HSPD1/780]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880780-500
  • BNC880780-100
  • Antigen symbol:
  • HSPD1
  • HSPD1
  • Antigen name:
  • HSP60
  • HSP60
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone HSPD1/780
  • Clone HSPD1/780
  • Reactivity:
  • Hamster, Pig, Rabbit, Monkey, Sheep, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Hamster, Pig, Rabbit, Monkey, Sheep, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 595053
  • 595053
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together