Anti-MLANA Mouse Monoclonal Antibody (CF568) [clone: HMB45 M2-7C10 M2-9E3 T311]
: Biotium
This antibody cocktail recognizes three melanoma-specific proteins, which include MART-1, Tyrosinase and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. This cocktail of three markers is designed for extremely sensitive labeling of formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.
Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: HMB45 M2-7C10 M2-9E3 T311
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
- Catalog No:
- BNC680701-500
- BNC680701-100
- Antigen symbol:
- MLANA
- MLANA
- Antigen name:
- Melanoma Marker
- Melanoma Marker
- Antigen synonyms:
- MLAN-A|Melanoma antigen recognized by T-cells 1 (MART-1)|TYR|PMEL17
- MLAN-A|Melanoma antigen recognized by T-cells 1 (MART-1)|TYR|PMEL17
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone HMB45 M2-7C10 M2-9E3 T311
- Clone HMB45 M2-7C10 M2-9E3 T311
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 154069-MART-1; 503555-Tyrosinase; 95972-gp100
- 154069-MART-1; 503555-Tyrosinase; 95972-gp100
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Frequently Bought Together