About this item
The c-Myc protein is a transcription factor, which is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies also have shown that c-Myc is essential for tumor cell development in vasculogenesis and angiogenesis that distribute blood throughout the cells, and which brought extensive attention in the development of new therapeutic approach for cancer treatment.
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: MYC699
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Gorilla, Human
Specifications
- Catalog No:
- BNC680699-500
- BNC680699-100
- Antigen symbol:
- MYC
- MYC
- Antigen name:
- c-Myc
- c-Myc
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone MYC699
- Clone MYC699
- Reactivity:
- Gorilla, Chimpanzee, Human
- Gorilla, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 202453
- 202453
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide