About this item
This antibody recognizes a protein of 54 kDa, which is identified as cyclin A. Its epitope is located amino acids 144-148 of human Cyclin A2. Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdk's. These cyclin/cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase. The D and E type cyclins regulate the passage of G1, while cyclin B is a critical regulator of mitosis. Mutation or disruption of normal cyclin A expression causes cells to arrest in G2-phase.
Type: Primary
Antigen: CCNA2
Clonality: Monoclonal
Clone: Clone E67
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Green monkey, Human, Mink, Monkey, Mouse
Specifications
- Catalog No:
- BNC400673-100
- BNC400673-500
- Antigen symbol:
- CCNA2
- CCNA2
- Antigen name:
- Cyclin A2
- Cyclin A2
- Antigen synonyms:
- Cyclin A2|CCN1|CCNA|CCNA2|CCNA2_HUMAN
- Cyclin A2|CCN1|CCNA|CCNA2|CCNA2_HUMAN
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone E67
- Clone E67
- Reactivity:
- Green monkey, Monkey, Human, Bovine, Mink, Mouse
- Green monkey, Monkey, Human, Bovine, Mink, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 58974
- 58974
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide