Order Entry
Belgium
ContactUsLinkComponent
Anti-TGFA Mouse Monoclonal Antibody (CF405S) [clone: MF9]
Anti-TGFA Mouse Monoclonal Antibody (CF405S) [clone: MF9]
Supplier:  Biotium
Certificates
Anti-TGFA Mouse Monoclonal Antibody (CF405S) [clone: MF9]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040644-100
  • BNC040644-500
  • Antigen symbol:
  • TGFA
  • TGFA
  • Antigen name:
  • TGFalpha
  • TGFalpha
  • Antigen synonyms:
  • TGFA|Waved 1|ETGF|TFGA|TGF Type 1|Wa1|EGF-like TGF
  • TGFA|Waved 1|ETGF|TFGA|TGF Type 1|Wa1|EGF-like TGF
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone MF9
  • Clone MF9
  • Reactivity:
  • Human, Bovine, Rat, Mouse
  • Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 170009
  • 170009
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together