Anti-PODXL Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 4F10]
BNUM0482-50
: Biotium
- Antibody type:Primary
- Antigen name:Podocalyxin
- Antigen symbol:PODX
- Clonality:Monoclonal
- Clone:Clone 4F10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Rabbit,Human,Rat
- Gene ID:732423
- Antigen synonyms:PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 4F10
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Rabbit, Rat