Anti-PODXL Mouse Monoclonal Antibody (Purified) [clone: 4F10]
Supplier: Biotium
Certificates
About this item
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 4F10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Rabbit, Rat
Specifications
- Catalog No:
- BNUB0482-500
- BNUB0482-100
- Antigen symbol:
- PODX
- PODX
- Antigen name:
- Podocalyxin
- Podocalyxin
- Antigen synonyms:
- PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
- PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone 4F10
- Clone 4F10
- Reactivity:
- Rabbit, Human, Rat
- Rabbit, Human, Rat
- Host:
- Mouse
- Mouse
- Gene ID:
- 732423
- 732423
- Isotype:
- IgM
- IgM
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
Specifications
Frequently Bought Together