About this item
This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium. Actin can be resolved on the basis of its isoelectric points into three distinctive components: alpha, beta, and gamma in order of increasing isoelectric point. Anti-muscle specific actin recognizes alpha and gamma isotypes of all muscle groups. Non-muscle cells such as vascular endothelial cells and connective tissues are non-reactive. Also, neoplastic cells of non-muscle-derived tissue such as carcinomas, melanomas, and lymphomas are negative.It stains tumors of smooth muscle (leiomyomas and leiomyosarcomas) as well as skeletal muscle (rhabdomyomas and rhabdomyosarcomas).
Type: Primary
Antigen: ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth)
Clonality: Monoclonal
Clone: HHF35
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cat, Chicken, Dog, Human, Mouse, Rabbit, Rat
Specifications
- Catalog No:
- BNC940445-500
- BNC940445-100
- Antigen symbol:
- ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth)
- ACTA1 (Skeletal), ACTA2 (Smooth), ACTG2 (Smooth)
- Antigen name:
- Muscle Specific Actin
- Muscle Specific Actin
- Antigen synonyms:
- Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
- Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone HHF35
- Clone HHF35
- Reactivity:
- Rabbit, Chicken, Human, Dog, Rat, Mouse, Cat
- Rabbit, Chicken, Human, Dog, Rat, Mouse, Cat
- Host:
- Mouse
- Mouse
- Gene ID:
- 118127, 1288 , 500483
- 118127, 1288 , 500483
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide