Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Testosterone Mouse Monoclonal Antibody (CF594) [clone: 4E1G2]
 :  Biotium
 
Anti-Testosterone Mouse Monoclonal Antibody (CF594) [clone: 4E1G2]
 :  Biotium

 

  • Catalog No:
  • BNC940250-100
  • BNC940250-500
  • Antigen name:
  • Testosterone
  • Testosterone
  • Antigen synonyms:
  • 4-Androsten-17-beta-ol-3-one|trans-Testosterone|17-beta-Hydroxy-4-androsten-3-one|17-beta-Hydroxy-3-oxo-4-androstene
  • 4-Androsten-17-beta-ol-3-one|trans-Testosterone|17-beta-Hydroxy-4-androsten-3-one|17-beta-Hydroxy-3-oxo-4-androstene
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone 4E1G2
  • Clone 4E1G2
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together