Order Entry
Belgium
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody (CF640R) [clone: C4/206]
Anti-CD4 Mouse Monoclonal Antibody (CF640R) [clone: C4/206]
Supplier:  Biotium
Certificates
Anti-CD4 Mouse Monoclonal Antibody (CF640R) [clone: C4/206]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400206-500
  • BNC400206-100
  • Antigen symbol:
  • CD4
  • CD4
  • Antigen synonyms:
  • L3T4|T cell differentiation antigen L3T4|Leu3|T-cell surface antigen T4/Leu-3|p55|T cell antigen T4/LEU3|Lymphocyte antigen CD4|Ly-4|T-cell surface glycoprotein CD4
  • L3T4|T cell differentiation antigen L3T4|Leu3|T-cell surface antigen T4/Leu-3|p55|T cell antigen T4/LEU3|Lymphocyte antigen CD4|Ly-4|T-cell surface glycoprotein CD4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone C4/206
  • Clone C4/206
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 631659
  • 631659
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together