Anti-PARP1 Mouse Monoclonal Antibody [clone: C-2-10]
Catalog # ENZOBMLSA2490050
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Poly (ADP-ribose) polymerase 1
- Antigen symbol:PARP1
- Clonality:Monoclonal
- Clone:C-2-10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Hamster,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:PARP1|PPOL|PARP-1|ADPRT1|pADPRT-1|PARP|ADPRT
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Purified calf thymus poly(ADP-ribose) polymerase (PARP)
- Purification:Purified by affinity chromatography
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: C-2-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Bovine