Anti-CASP9 Rabbit Polyclonal Antibody
Catalog # BMOLSA321-0100
Supplier: BIOMOL RESEARCH LABORATORIES
Specifications
- Antibody type:Primary
- Antigen name:Caspase 9
- Antigen symbol:CASP9
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Liquid. Neat serum containing sodium azide.
- Antigen synonyms:EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human