Order Entry
Belgium
ContactUsLinkComponent
 
DNA extraction kits, tissue DNA extraction kit, MPure™, MP Biomedicals
  ICNA117022400
 :  
undefined
DNA extraction kits, tissue DNA extraction kit, MPure™, MP Biomedicals
  ICNA117022400
 :  

 

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    MPure™ Tissue DNA Extraction Kit, 48 tests
  • Description:
    MPure™ Tissue DNA Extraction Kit, 48 tests

 

 

MPure™ Tissue DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for extraction and purification of genomic DNA from a variety of animal tissues, swab samples and blood stains. Nucleic acids extracted and purified from MPure™ Tissue DNA Extraction Kits can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.