Order Entry
Belgium
ContactUsLinkComponent
 
VWR® ARISTAR®, Zirconium standard solutions for ICP-MS
  85649.180
 :  
undefined
VWR® ARISTAR®, Zirconium standard solutions for ICP-MS
  85649.180
 :  

 

  • Concentration:
    10 ppm
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    151
  • Element:
    Zirconium
  • Symbol:
    Zr

 

 

  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
 : Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities