VWR® ARISTAR®, Ytterbium Standard Solutions for ICP-MS
85647.180
:
- Concentration:10 ppm
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:144
- Element:Ytterbium
- Symbol:Yb
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities