VWR® ARISTAR®, Terbium Standard Solutions for ICP-MS
Catalog # 85633.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:10 ppm
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:130
- Element:Terbium
- Symbol:Tb
Specifications
About this item
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0.5% (typical)
- Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities