Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® ARISTAR®, Selenium Standard Solutions for ICP-MS

VWR® ARISTAR®, Selenium Standard Solutions for ICP-MS

VWR® ARISTAR®, Selenium Standard Solutions for ICP-MS
Supplier:  VWR Chemicals
Certificates
VWR® ARISTAR®, Selenium Standard Solutions for ICP-MS
VWR® ARISTAR®, Selenium Standard Solutions for ICP-MS

2 Options Available Below

Product Details & Documents
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis

Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities

Product Family Options

Product Information

  • Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Selenium
    Se
    10 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 131,00 €
    131,00 €
    1 * 100 mL
       131,00 €

  • Selenium
    Se
    1000 mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.