Order Entry
Belgium
ContactUsLinkComponent
 
Nucleic acid purification system, MPure-12™, MP Biomedicals
  ICNA117002200
Nucleic acid purification system, MPure-12™, MP Biomedicals
  ICNA117002200

 

  • Description:
    MPure-12™ Nucleic Acid Purification System
  • Pk:
    1

 

 

MPure-12™ is a bench top automated system for rapid purification of nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology. It has a uniquely designed magnetic bead processing chamber and is fully integrated with easy to use pre-packaged reagent kits.

  • Fully automated and integrated platform that offers true walk-away operation
  • Purify nucleic acids of the highest yield and purity, ready for immediate use in downstream applications
  • Flexible in throughput, process 1 to 12 samples from a wide variety of biological specimens in a single run
  • Uniquely designed Polygon Reaction Chamber minimises cross contamination and mag bead carryover
  • Simple operation, Bar-code Scanner quickly enters program parameters in seconds
  • Minimize nucleic acid loss and degradation
  • Reproducible, sample-to-sample consistency, scalable and convenient
  • Closed system that offers increased safety and efficiency

MPure-12™ is a state of the art bench-top automated nucleic acid purification system that rapidly provides superior yield and purity of nucleic acids for any downstream application including qPCR, RE digestion and Southerns. The MPure-12 system utilizes a unique magnetic bead separation technology, is fully integrated with easy to use pre-packaged reagent kits and provides superior nucleic acid purification results from a wide variety of biological specimens.

Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.

Specifications
Sample Type: Plasma, Serum, Blood, Plants, Cultured Cells, Bacteria, FFPE, Forensic samples, Swabs, Tissues
Capacity: Purify Nucleic acids from 100 to 1000 µl of sample
Sensitivity: As little as 5 IU spiked (about 1 IU in PCR reaction) sample can be detected, proving the excellent sensitivity and linearity of isolation
Time setting: 30 to 70 minutes
Dimensions: 48W×70D×52H cm
Weight: 43 kg
Power requirements: 110 to 240 V, 50/60 Hz

 : Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Reagent kits